missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pref-1/DLK1/FA1, Mouse anti-Human, Clone: D1L165-6, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80048-50UG
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Pref-1/DLK1/FA1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8788 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| D1L165-6 | |
| Unconjugated | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| DLK1 | |
| Recombinant human Pref-1/DLK1/FA1. | |
| 50 μg | |
| Primary | |
| Recognizes human Pref-1/DLK1/FA1. Detects a band of ∽68kDa by Western blot. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction