missing translation for 'onlineSavingsMsg'
Learn More

Prednisone, TRC

Product Code. 17984075
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17984075 25 mg 25mg
17984885 100 mg 100mg
17953835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17984075 Supplier Toronto Research Chemicals Supplier No. TRCP70378025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-03-2
Molecular Formula C21 H26 O5
Molecular Weight (g/mol) 358.43
Synonym Prednisone, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione, Prednisolone Imp. B (EP), 1,2-Dehydrocortisone, 1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Dehydrocortisone, 17,21-Dihydroxypregn-1,4-diene-3,11,20-trione, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Adasone, Ancortone, Apo-Prednisone, Bicortone, Cartancyl, Colisone, Cordrol, Cortan, Cortidelt, Dacorten, Dacortin, Decortancyl, Decortin, Decortisyl, Dehydrocortisone, Dekortin, Delcortin, Dellacort, Dellacort A, Delta E, Delta-Cortelan, Delta-Dome, Deltacortene, Deltacortisone, Deltacortone, Deltasone, Deltison, Deltisona, Deltisone, Deltra, Di-Adreson, Drazone, Econosone, Encorton, Encortone, Enkorton, Fernisone, Hostacortin, Liquid Pred, Me-Korti, Metacortandracin, Meticorten, NSC 10023, Nisona, Nurison, Orasone, Panafcort, Panasol, Paracort, Parmenison, Pehacort, Prednicen M, Prednicorm, Prednicort, Prednicot, Prednidib, Prednilonga, Predniment, Prednison, Prednitone, Pronison, Pulmison, Rectodelt, Retrocortine, Sone, Sterapred, Supercortil, Ultracorten, Ultracortene, Uniplen, Winpred, δ-Cortisone, δ1-Cortisone, δ1-Dehydrocortisone
IUPAC Name (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
SMILES C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1
Formula Weight 358.178
Percent Purity >95%
Chemical Name or Material Prednisone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.