missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone, Dr. Ehrenstorfer

Product Code. 30316645
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316645 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316645 Supplier Dr. Ehrenstorfer Supplier No. DREC16286500100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednisolone
CAS 50-24-8
Analyte or Component Names Prednisolone
Shipping Condition +20°C
Molecular Formula C21 H28 O5
Certifications/Compliance ISO 17034
Synonym Hydrocortisone Imp. A (EP), Prednisolone Acetate Imp. B (EP), 11,17,21-Trihydroxy-(11beta)-Pregna-1,4-diene-3,20-dione, 11Beta,17,21-trihydroxy-Pregna-1,4-diene-3,20-dione, Delta1-Cortisol, Delta1-Dehydrocortisol, Delta1-Dehydrohydrocortisone, Delta1-Hydrocortisone, 1,2-Dehydrohydrocortisone, 1,4-Pregnadiene-11beta,17alpha,21-triol-3,20-dione, 1,4-Pregnadiene-3,20-dione-11beta,17alpha,21-triol, 1-Dehydrohydrocortisone, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 11beta,17alpha,21-Trihydroxypregna-1,4-diene-3,20-dione, Co-Hydeltra, Codelcortone, Cortalone, Decaprednil, Decortin H, Delcortol, Delta F, Delta-Cortef, Delta-Ef-Cortelan, Delta-stab, Deltacortenol, Deltacortril, Deltacortril Enteric, Deltahydrocortisone, Deltasolone, Deltisilone, Di-Adreson F, Dicortol, Donisolone, Eazolin D, Fernisolone, Flamasone, Hostacortin H, Hydeltra, Hydeltrone, Hydrodeltalone, Hydrodeltisone, Hydroretrocortin, Hydroretrocortine, Klismacort, Metacortandralone, Meti-Derm, Meticortelone, NSC 9120, NSC 9900, Panafcortelone, Paracortol, Precortalon, Precortancyl, Precortilon, Precortisyl, Predne-Dome, Prednelan, Predni-Dome, Prednicen, Predniretard, Prednisolone, Predonin, Predonine, Preflam, Prelone, Prenolone, Scherisolon, Solone, Steran, Sterane, Sterolone, Supercortisol, Ulacort, Ultracorten H, Ultracortene H Prednisolone Acetate Imp. B (EP), Hydrocortisone Impurity A, Methylprednisolone Impurity K, Prednicarbate Impurity A, Prednisolone Acetate Impurity B, Prednisone Impurity B
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 360.44
Quantity 100 mg
Formula Weight 360.1937 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.