missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednisolone acetate, MedChemExpress
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Description
Prednisolone acetate (Prednisolone 21-acetate) is an adrenal cortico hormones, with anti-inflammatory, anti-allergic and immune suppressive effects.
Specifications
Specifications
| Chemical Name or Material | Prednisolone acetate |
| CAS | 52-21-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H30O6 |
| Quantity | 500 mg |
| Synonym | Prednisolone 21-acetate |
| Solubility Information | DMSO : 50 mg/mL (124.23 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.66 mM; Need ultrasonic) |
| SMILES | C[C@@]12[C@](C(COC(C)=O)=O)(O)CC[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O |
| Molecular Weight (g/mol) | 402.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction