missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone (1.0 mg/mL in Methanol), TRC

Product Code. 30466525
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466525 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466525 Supplier Toronto Research Chemicals Supplier No. TRCKIT825510X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 50-24-8
Molecular Formula C21 H28 O5
Quantity 10 x 1 mL
Synonym Prednisolone Acetate Imp. B (EP), Hydrocortisone Imp. A (EP), Pregna-1,4-diene-3,20-dione, 11,17,21-trihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy- (8CI), (11β)-11,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 1,2-Dehydrohydrocortisone, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1,4-Pregnadiene-3,20-dione-11β,17α,21-triol, 1-Dehydrohydrocortisone, 11β,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 11β,17α,21-Trihydroxypregna-1,4-diene-3,20-dione, Aprednislon, Co-Hydeltra, Codelcortone, Cortalone, Decaprednil, Decortin H, Delcortol, Delta F, Delta-Cortef, Delta-Ef-Cortelan, Delta-stab, Deltacortenol, Deltacortril, Deltacortril Enteric, Deltahydrocortisone, Deltasolone, Deltisilone, Di-Adreson F, Dicortol, Donisolone, Eazolin D, Fernisolone, Flamasone, Hostacortin H, Hydeltra, Hydeltrone, Hydrodeltalone, Hydrodeltisone, Hydroretrocortin, Hydroretrocortine, Klismacort, Metacortandralone, Meti-Derm, Meticortelone, Millipred, NSC 9120, NSC 9900, Panafcortelone, Paracortol, Precortalon, Precortancyl, Precortilon, Precortisyl, Predne-Dome, Prednelan, Predni-Dome, Prednicare, Prednicen, Predniretard, Prednisolona Syntex, Prednisolone, Predonin, Predonine, Preflam, Prelone, Prenolone, Scherisolon, Solone, Steran, Sterane, Sterolone, Supercortisol, Ulacort, Ultracorten H, Ultracortene H, Vetsolone, δ1-Cortisol, δ1-Dehydrocortisol, δ1-Dehydrohydrocortisone, δ1-Hydrocortisone
InChI Formula InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 360.44
Formula Weight 360.1937
Concentration or Composition (by Analyte or Components) Prednisolone 1 mg/mL
Chemical Name or Material Prednisolone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.