missing translation for 'onlineSavingsMsg'
Learn More

Prednienic Acid, TRC

Product Code. 30134215
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134215 5 g 5g
17992195 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134215 Supplier Toronto Research Chemicals Supplier No. TRCP7037105G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37927-29-0
Molecular Formula C20 H26 O5
Molecular Weight (g/mol) 346.42
Synonym Prednienic Acid, Prednisolone Impurity B, (11β,17α)-11,17-Dihydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid;
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-17-carboxylic acid
SMILES C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H26O5/c1-18-7-5-12(21)9-11(18)3-4-13-14-6-8-20(25,17(23)24)19(14,2)10-15(22)16(13)18/h5,7,9,13-16,22,25H,3-4,6,8,10H2,1-2H3,(H,23,24)/t13-,14-,15-,16+,18-,19-,20-/m0/s1
Formula Weight 346.178
Chemical Name or Material Prednienic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.