missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Precision Plate Holder for 7500 Real-Time PCR Systems

Product Code. 17115324 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17115324 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17115324 Supplier Applied Biosystems™ Supplier No. A24820
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Required for use of MicroAmp™ EnduraPlate™ optical reaction plates on Applied Biosystems™ 7500 real-time PCR systems

  • This precision plate holder is required for use of MicroAmp™ EnduraPlate™ optical reaction plates on Applied Biosystems™ 7500 real-time PCR systems. It also acommodates MicroAmp™ optical reaction plates
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 7500 Reactions
For Use With (Equipment) 7500 System
Product Type Precision Plate Holder
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 Ea.
PCR Method qPCR

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.