Learn More
MakerBot™ Precision Material PVA
Product Code. 16331556
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
MakerBot Method
Product Code. For Use With (Equipment)
16331556 MakerBot Method
1 options
Product Code. 16331556 Supplier MakerBot™ Supplier No. MKS000098
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MakerBot™ Precision Material PVA for the Method 3D Printer is easy to use and ideal for early-stage concept development.
  • Smart Spool™ provides valuable information including material type, color, and amount remaining via an RFID chip directly to MakerBot Print, while a desiccant in the spool maintains a low moisture level inside the drawer bay
  • Glass Transition Temperature: 60°C
  • Melting Temperature: 170°C
  • Extruder Temperature: 210°C
Specifications
For Use With (Equipment) MakerBot Method
Product Type Precision Material
Material PVA