missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PrecisION™ hKv1.8 Recombinant Stable Cell Line
Description
The PrecisION™ hKv1.8 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Kv1.8 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Kv1.8 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Ion Channel Activity |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Quantity | 2 Vials |
| Target Entry | Kv1.8 |
| Accession Number | NM_005549 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?