Learn More
Thermo Scientific™ Precise™ 12%, Tris-Glycine, 5.0 mm, 10-well
Separation Range: 14 to 205 kd; Separation Type: Denaturing, Native; For Use With: Novex XCell I and II; Novex XCell II Surelock; IBI Scientific Universal Protein Systems
Brand: Thermo Scientific™ 25262
Code : PN
Additional Details : CAS Number : 79-06-1 Weight : 0.01000kg
Description
Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.
Precise Tris-Glycine Gels are cast in durable 10 × 8cm plastic cassettes to fit in Mini-PROTEAN™ gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 × 8cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.Highlights:
High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands
Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels
Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains
Compatible – these 10 x 8cm cassettes fit Bio-Rad™ Mini-PROTEAN™ and Hoefer™ Mighty Small II gel tanks, as well as other similar tanks
Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination
Specifications:
Cassette Dimensions: 10cm x 8.0cm x 5mm
Gel Dimensions: 8cm x 6.8cm x 1mm
Storage Conditions: 4°C
Shelf Life: 12 months from date of purchase
Stacking Gel: 4%
Buffer System in Gel: Tris-HCl, pH 7
SDS: None
Required Running Buffer: Tris-glycine-SDS
Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS
Gel Tank Compatibility (10cm x 8cm x 5mm): Bio-Rad Mini-PROTEAN II and 3; Bio-Rad Mini-PROTEAN Tetra Cell; Hoefer Tall Mighty Small (SE 280); Hoefer Mighty Small II (SE 260); IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 10cm Gels
Specifications
12% Tris-Glycine Gels, 10 x 8cm, 10-well | |
12% | |
Tris-Glycine | |
14 to 205 kDa | |
10-well | |
10/Pk. | |
Standard | |
8 cm |
Novex XCell I and II, Novex XCell II Surelock, IBI Scientific Universal Protein Systems | |
0.5 cm | |
Homogeneous Page | |
Denaturing, Native | |
Precast 12% polyacrylamide gels for SDS-PAGE | |
10 samples per gel, 50uL capacity per well | |
10 cm |