missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precise™ 12%, Tris-HEPES, Protein Gel, 10-well

Product Code. 11861385 Shop All Thermo Scientific Products
Click to view available options
Wells:
10-well
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11861385

Brand: Thermo Scientific™ 25202

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Separation Range: 14 to 205 kd

Thermo Scientific™ Precise Tris-HEPES Gels are precast polyacrylamide gels for protein electrophoresis (PAGE) that use Tris-HEPES running buffer, have long shelf lives, and provide compatibility with different commercial gel tanks.

Precise Gels are cast in a durable plastic cassette using a neutral-pH buffer that prevents polyacrylamide breakdown and results in a long shelf life. High-resolution staining and transfer of proteins is accomplished quickly on these 1mm thick mini gels. Gels are individually packaged in an easy-to-open plastic pouch and are ready to run with no comb or tape to remove. Following electrophoresis, the cassettes are easy to dismantle (no special tools required) without damaging the gel. Precise Gels are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well, 12-well and 15-well formats.

Highlights:

  • High-performance – unique running buffer produces excellent separation and high-resolution protein bands that stain easily using coomassie and silver stains

  • Long shelf life – guaranteed 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes in just 90 minutes

  • Large-volume wells – innovative well design accommodates twice the sample volume of typical mini-gel cassette wells (10-well=50μL, 12-well=30μL; 15-well=25μL)

  • Compatible – cassettes fit many popular mini-gel tanks, and gels can be loaded using standard Laemmli sample loading buffers

  • Several convenient styles – choose from five gel densities and three well formats

Additional Features:

  • Cassette dimensions: 10cm × 8.5cm × 4.5mm
  • Gel dimensions: 8cm × 5.8cm × 1mm
  • Storage conditions: 1 year at 4°C
  • Stacking gel: 4% polyacrylamide
  • Buffer system in gel: Tris-HCl, pH 7
  • SDS content: none
  • Required running buffer: Tris-HEPES-SDS (Part No. 28398)
  • Recommended sample buffer: Tris-HCl-SDS (any type)
Precise Protein Gels are compatible with the following gel tanks: BioRad™ Mini-PROTEAN™ II and 3; Life Technologies XCell I, II™ and Surelock™; IBI Universal Protein System; EC 4-Cell; Hoefer Tall Mighty Small™ (SE 280); Hoefer Mighty Small™ II (SE 260/ SE 250); Hoefer Mini VE System (SE 300); C.B.S. Scientific MGV 302/402; Daiichi Mini 2-Gel and 6-Gel; GradiGel Mini 4-Cell; Fisher Biotech Vertical Minigel System.
TRUSTED_SUSTAINABILITY

Specifications

Description 12% Tris-HEPES Gels, 10 x 8.5cm, 10-Well
Gel Percentage 12%
Gel Thickness 4.5 mm
Gel Type Tris-HEPES
Product Type Homogeneous Page
Separation of Protein
Separation Range 14 to 205 kDa
Wells 10-well
Formulation Precast 12% polyacrylamide gels for SDS-PAGE
Quantity 10/Pk.
Sufficient For 10 samples per gel, 50uL capacity per well
Well Design Standard
Length (Metric) 100 mm
Width (Metric) 85 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.