Learn More
Thermo Scientific™ Precise™ 10%, Tris-HEPES, Protein Gel, 12-well
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Precise Tris-HEPES Gels are precast polyacrylamide gels for protein electrophoresis (PAGE) that use Tris-HEPES running buffer, have long shelf lives, and provide compatibility with different commercial gel tanks.
Highlights:
High-performance – unique running buffer produces excellent separation and high-resolution protein bands that stain easily using coomassie and silver stains
Long shelf life – guaranteed 12-month stability ensures consistent performance for the duration of each package of gels
Fast – 45-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes in just 90 minutes
Large-volume wells – innovative well design accommodates twice the sample volume of typical mini-gel cassette wells (10-well=50μL, 12-well=30μL; 15-well=25μL)
Compatible – cassettes fit many popular mini-gel tanks, and gels can be loaded using standard Laemmli sample loading buffers
Several convenient styles – choose from five gel densities and three well formats
Additional Features:
- Cassette dimensions: 10cm × 8.5cm × 4.5mm
- Gel dimensions: 8cm × 5.8cm × 1mm
- Storage conditions: 1 year at 4°C
- Stacking gel: 4% polyacrylamide
- Buffer system in gel: Tris-HCl, pH 7
- SDS content: none
- Required running buffer: Tris-HEPES-SDS (Part No. 28398)
- Recommended sample buffer: Tris-HCl-SDS (any type)
Specifications
Specifications
| Description | 10% Tris-HEPES Gels, 10 x 8.5cm, 12-Well |
| Gel Percentage | 10% |
| Gel Thickness | 4.5 mm |
| Gel Type | Tris-HEPES |
| Separation of | Protein |
| Separation Range | 24 to 205 kDa |
| Wells | 12-well |
| Formulation | Precast 10% polyacrylamide gels for SDS-PAGE |
| Quantity | 10/Pk. |
| Sufficient For | 12 samples per gel, 30uL capacity per well |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.