missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precise™ 12%, Tris-Glycine, 7.0 mm, 12-well

Product Code. 11864594 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11864594

Brand: Thermo Scientific™ 25252

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Separation Range: 14 to 205 kd; Separation Type: Denaturing, Native; For Use With: Novex XCell I and II; Novex XCell II Surelock; IBI Scientific Universal Protein Systems

Thermo Scientific™ Precise Tris-Glycine Gels are long shelf-life, fast run-time polyacrylamide gels for SDS-PAGE that use the popular Laemmli buffer system and commercial gel tanks to provide outstanding protein resolution and separation.

Precise Tris-Glycine Gels are cast in durable 10 x 10cm plastic cassettes to fit in Novex™ XCell gel tanks. The gels are individually packaged in easy-to-open plastic pouches and are ready for electrophoresis with no comb or tape to remove. Following electrophoresis, the easy-open cassette design allows gels to be removed damage-free without special tools. Precise Gels in the 10 x 10cm size are available in gradient (4 to 20% and 8 to 16%) and fixed (8%, 10% and 12%) concentrations and in 10-well and 12-well formats.

Highlights:

  • High-performance – Tris-glycine running buffer produces excellent separation and high-resolution protein bands

  • Long shelf-life – up to 12-month stability ensures consistent performance for the duration of each package of gels

  • Fast – 60-minute run-time provides results quickly; transfers efficiently to nitrocellulose and PVDF membranes; stains quickly with coomassie and silver stains

  • Compatible – these 10 x 10cm cassettes fit the Novex™ XCell and a other similar tanks

  • Easy to load – sample wells reinforced with plastic to eliminate damage when loading and well-to-well contamination

Specifications:

  • Cassette Dimensions: 10cm × 10cm × 7mm

  • Gel Dimensions: 8cm × 8.8cm × 1mm

  • Storage Conditions: 4°C

  • Shelf Life: 12 months from date of purchase

  • Stacking Gel: 4%

  • Buffer System in Gel: Tris-HCl, pH 7

  • SDS: None

  • Required Running Buffer: Tris-glycine-SDS

  • Recommended Sample Buffer: Tris-HCl-SDS or LDS-SDS

  • Gel Tank CompatibilityCompatibility (10cm x 10cm x 7mm): Novex XCell I and II; Novex XCell II™ Surelock™; IBI Scientific Universal Protein Systems; For gels that are compatible with other tanks, see our 10 x 8cm Gels

TRUSTED_SUSTAINABILITY

Specifications

Description 12% Tris-Glycine Gels, 10 x 10cm, 12-well
For Use With (Equipment) Novex XCell I and II, Novex XCell II Surelock, IBI Scientific Universal Protein Systems
Gel Percentage 12%
Gel Thickness 0.7 cm
Gel Type Tris-Glycine
Product Type Homogeneous Page
Separation Range 14 to 205 kDa
Separation Type Denaturing, Native
Wells 12-well
Formulation Precast 12% polyacrylamide gels for SDS-PAGE
Quantity 10/Pk.
Sufficient For 12 samples per gel, 30uL capacity per well
Well Design Standard
Length (Metric) 10 cm
Width (Metric) 10 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.