Learn More
Precious Metals, plasma standard solution, Specpure™, Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 014652.AE
137.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
Precious Metals, plasma standard solution is useful as a standard for the determination of metals by inductively coupled plasma-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Precious Metals, plasma standard solution | |
Au, Ir, Os, Pd, Pt, Re, Rh, Ru @ 100μg/mL | |
MFCD00151264 | |
Specpure™ | |
Liquid |
Matrix: 20% HCl | |
100 mL | |
UN1789 | |
Miscible with water |
RUO – Research Use Only