missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ PrEBM™ Prostate Epithelial Cell Growth Basal Medium
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
- Basal medium.
- Can be used with CC-4177 (prostate epithelial cell growth medium SingleQuots™ Supplements and Growth Factors).
- Full PrEGM™ BulletKit™ consisting of basal medium and supplements can be purchased together (CC-3166).
- Specially formulated for the maintenance and expansion of human prostate epithelial cells.
- Serum free
Specifications
Specifications
| Cell Type | Epithelial Cells |
| Product Type | Basal Medium |
| Quantity | 500 mL |
| pH | 7.4 to 7.6 |
| Color | Red |
| Content And Storage | Store in dark at 2 to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction