missing translation for 'onlineSavingsMsg'
Learn More

PREB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB120-3422-0.025ml

 View more versions of this product

Product Code. 18584673

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

PREB Polyclonal specifically detects PREB in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PREB
Polyclonal
Western Blot 1:500
MGC3467, prolactin regulatory binding-element protein, prolactin regulatory element binding, prolactin regulatory element-binding protein, SEC12Mammalian guanine nucleotide exchange factor mSec12
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Detects recombinant mouse PREB (Prolactin Regulatory Element Binding protein).
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
PREB
Recombinant mouse PREB.
0.025 mL
Transcription Factors and Regulators
10113
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only