missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pre-Weigh Buffered Peptone Water (BPW) (ISO) Agar

Product Code. p-6810172
Click to view available options
Quantity:
10 x Sachets for 8 L Medium
20 x Sachets for 1 L Medium
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15882477

Brand: Thermo Scientific™ PW10498L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Speed up your workflow when preparing medium for pre-enrichment of Salmonella species from foods with pre-weighed media sachets.

Speed up your workflow when preparing microbial culture medium for pre-enrichment of Salmonella species from food samples with Thermo Scientific™ Pre-weigh Buffered Peptone Water (ISO) Agar. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.

Pre-Weigh Buffered Peptone Water (ISO) sachets contain the familiar Thermo Scientific Oxoid Buffered Peptone Water (ISO) medium (Manf. Part No. CM1049) and are part of a range of our most frequently used and well-characterized dehydrated culture media, which is now available in handy pre-weighed foil packages.

Simply tear open the selected media sachet, add to the required volume of water, mix, and autoclave as usual.

  • Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
  • Convenient: rip and tip sachets enable faster media production and simplified workflows
  • Compliant: proven and validated media formats deliver confidence and reliable performance
  • Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Pre-Weigh Buffered Peptone Water (ISO)
Form Powder
Product Type Agar
Quantity 10 x Sachets for 8 L Medium

For Laboratory Use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.