missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pre-supplemented Dichloran-Glycerol 18% (DG18) (ISO) Agar Base (Dehydrated)
Click to view available options
Quantity:
500 g
Unit Size:
Each
Description
Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Pre-supplemented Dichloran-Glycerol 18% (DG18) (ISO) Agar Base (Dehydrated) while adhering to BS ISO 21527-2: 2008. Due to the presence of antibiotic Chloramphenicol in the formulation, the medium provides selectivity against bacteria as well as reducing harmful chemical handling in the laboratory.
Use Pre-supplemented Dichloran-Glycerol 18% (DG18) (ISO) Agar Base for the enumeration and isolation of xenophilic molds from dried and semi-dried foods like spices, dried fruits, confectionery, cereals, nuts, dried meat, and fish products.
The formulation contains glycerol at 18% (w/w) which lowers the water activity (aW) from 0.999 to 0.95.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
- Adheres to BS ISO 21527-2: 2008 standards.
- Makes colony counting easier: contains antifungal agent Dichloran which inhibits spreading of mucoraceous fungi and restricts the colony size of other genera.
The formulation contains glycerol at 18% (w/w) which lowers the water activity (aW) from 0.999 to 0.95.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Description | Pre-supplemented Dichloran-Glycerol 18% (DG18) (ISO) Agar Base (Dehydrated) |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
Safety and Handling
Product Identifier
- Pre Supplemented DG18 (ISO) Agar Base
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Citric acid, Chloramphenicol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction