Learn More
LTE Scientific™ Pre/Post Vacuum System with Steam Jacket for the TC/360/KE1 Model
Click to view available options
For Use With (Equipment):
TC/360/KE1 Option
Description
- Pre/Post Vacuum System with Steam Jacket
- Vacuum helps speed up cycle times
- Vacuum helps eliminate air pockets within the chamber
- Vacuum assists with the drying cycle
- Steam jacket assists with the drying cycle
Specifications
Specifications
| For Use With (Equipment) | TC/360/KE1 Option |
| Model | TC/360/KE5 |
| Sterility | Non-sterile |