missing translation for 'onlineSavingsMsg'
Learn More

pre-mRNA cleavage factor I (59 kDa subunit) Antibody, Novus Biologicals™

Product Code. 18433279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433279 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433279 Supplier Novus Biologicals Supplier No. H00079869B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

pre-mRNA cleavage factor I (59 kDa subunit) Polyclonal antibody specifically detects pre-mRNA cleavage factor I (59 kDa subunit) in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen pre-mRNA cleavage factor I (59 kDa subunit)
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_079087.2
Gene Alias CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, CPSF 59 kDa subunit, FLJ12529,59 kDa subunit, FLJ39024, MGC9315, pre-mRNA cleavage factor I, 59 kDa subunit
Host Species Mouse
Immunogen FLJ12529 (NP_079087.2, 1 a.a. - 471 a.a.) full-length human protein. MSEGVDLIDIYADEEFNQDPEFNNTDQIDLYDDVLTATSQPSDDRSSSTEPPPPVRQEPSPKPNNKTPAILYTYSGLRNRRAAVYVGSFSWWTTDQQLIQVIRSIGVYDVVELKFAENRANGQSKGYAEVVVASENSVHKLLELLPGKVLNGEKVDVRPATRQNLSQFEAQARKRECVRVPRGGIPPRAHSRDSSDSADGRATPSENLVPSSARVDKPPSVLPYFNRPPSALPLMGLPPPPIPPPPPLSSSFGVPPPPPGIHYQHLMPPPPRLPPHLAVPPPGAIPPALHLNPAFFPPPNATVGPPPDTYMKASAPYNHHGSRDSGPPPSTVSEAEFEDIMKRNRAISSSAISKAVSGASAGDYSDAIETLLTAIAVIKQSRVANDERCRVLISSLKDCLHGIEAKSYSVGASGSSSRKRHRSRERSPSRSRESSRRHRDLLHNEDRHDDYFQERNREHERHRDRERDRHH
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79869
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.