missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pre-mRNA cleavage factor I (59 kDa subunit) Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61600R
This item is not returnable.
View return policy
Description
pre-mRNA cleavage factor I (59 kDa subunit) Polyclonal antibody specifically detects pre-mRNA cleavage factor I (59 kDa subunit) in Human, Mammalia, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| pre-mRNA cleavage factor I (59 kDa subunit) | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, CPSF 59 kDa subunit, FLJ12529,59 kDa subunit, FLJ39024, MGC9315, pre-mRNA cleavage factor I, 59 kDa subunit | |
| The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human cleavage and polyadenylation specificity factor 7, 59 kDa subunit using the numbering given in entry NP_079087.2 (GeneID 79869). | |
| 0.1 mL | |
| Primary | |
| Human, Mammalia, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79869 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction