Learn More
3M™ Pre-filter Pads
Extends the life of your Powered Air Respirator by capturing large particles that can clog a filter.
£28.30
Specifications
Product Type Prefilter Pad
View More Specs
Products 1
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
12374669
View Documents Promotion Details
3M™
7000006600
461-00-02P24X5
£28.30
Pack of 24
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
  • Frequent changing of prefilters can optimize performance
  • For use with 3M Jupiter™ Powered Air Respirators
Specifications
Specifications
Prefilter Pad