missing translation for 'onlineSavingsMsg'
Learn More

Solotec Scientific Limited Waysafe™ G4 Pre-Filter For Use With Waysafe™ SA and GP Enclosures

Product Code. 12797296
Change view
Click to view available options
For Use With (Equipment):
Waysafe SA and GP Enclosures
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12797296 Waysafe SA and GP Enclosures Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12797296 Supplier Solotec Scientific Limited Supplier No. 300.G4

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

G4 Prefilter pads for Waysafe GP and SA enclosures (also WS2, WS3, & Mediaway) (pack of 10).

Replacement pre-filters, for use with Waysafe™ SA and GP enclosures (pack of 10)

Specifications

Filter Type G4 Prefilter
For Use With (Equipment) Waysafe SA and GP Enclosures
Weight (Metric) 2 kg
Height (Metric) 5 cm
Dimensions 32 x 32 x 5 cm
Quantity 10 Pk.
Depth (Metric) 32 cm
Description G4 Prefilters
Length (Metric) 32 cm
Material Thermal Bonded 100% Polyester Fiber
For Use With (Application) Large Particulate Filtration
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.