missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Pre-drilled Rubber Stopper Without Holes

Product Code. 11714538
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11714538 Blue Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11714538 Supplier Saint Gobain Life Sciences Supplier No. 20304120

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use these stoppers for a variety of applications. Saint Gobain Life Sciences Rubber Stoppers Without Holes are ideal for chemical transfer.

  • Designed to keep container contents secure
  • Ideal for securing glass, metal and plastic containers
  • Easy to use
TRUSTED_SUSTAINABILITY

Specifications

Height (Metric) 35mm
Quantity 20/Pk.
Color Blue
Diameter (Metric) Bottom 34 mm
Diameter (Metric) Top 41 mm
Material Natural Rubber
Product Type Rubber Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.