missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pre-cut Blank Labels for BBP3X, S3XXX, and i3300 Printers

Product Code. 15971001
Click to view available options
Color:
White
Yellow
Height (Metric):
72 mm
75 mm
152 mm
Width (Metric):
28.6 mm
57 mm
101 mm
Vertical Repeat (Metric):
82.6 mm
158.8 mm
Unit Size:
Pack of 175
Pack of 300
This item is not returnable. View return policy

Product Code. 15971001

Brand: Brady™ 120930

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The material of these labels is ideal for indoor and outdoor facility labels.

  • Ideal for 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
  • Vinyl material

Specifications

Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
Material Vinyl (B-7569)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labeling
For Use With (Equipment) BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, S3000, S3100, and i5300 Industrial Label Printers
Labels per Row 1
Quantity 175 Labels
Shape Rectangle
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 158.8 mm
Width (Metric) 101 mm
Height (Metric) 152 mm
Alternate Part Number B30-25-7569-WT
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.