missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pre-BCR Rat anti-Mouse, BUV615, Clone: SL156, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 752350
This item is not returnable.
View return policy
Description
Pre-BCR Monoclonal antibody specifically detects Pre-BCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Pre-BCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms Pre-B Cell Receptor BUV615 SL156 50 μg | |
| Purified soluble complex of recombinant μ IgH, λ5, and VpreB | |
| RUO | |
| Pre-B Cell Receptor | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| SL156 | |
| Brilliant Ultraviolet 615 | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction