Learn More
PRDX6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PRDX6 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18050506
|
Proteintech
CL488-13585-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074785
|
Proteintech
CL488-13585-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PRDX6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRDX6. This antibody recognizes human, mouse, rat antigen. The PRDX6 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| PRDX6 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| 1 Cys, 1 Cys peroxiredoxin, 1 Cys PRX, 24 kDa protein, aiPLA2, Antioxidant protein 2, AOP2, KIAA0106, Liver 2D page spot 40, NSGPx, p29, peroxiredoxin 6, PRDX6, PRX, Red blood cells page spot 12 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 9588 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.