Learn More
Invitrogen™ PRDX3 Recombinant Rabbit Monoclonal Antibody (24GB935)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557561
 View more versions of this product
Product Code. 30276122
  • £172.00 / 20µL
Description
PRDX3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRDX3
Recombinant Monoclonal
Unconjugated
PRDX3
antioxidant protein; antioxidant protein 1; anti-oxidant protein 1; Aop1; AOP-1; AW822249; D0Tohi1; Ef2l; HBC189; Mer5; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; perioredoxin-3; peroxiredoxin 3; peroxiredoxin III; Peroxiredoxin-3; Prdx3; PRO1748; Protein MER5; Protein MER5 homolog; Prx; Prx 3; PRx III; Prx3; PRX-3; Prx-III; SP22; SP-22; TDXM; thioredoxin-dependent peroxide reductase, mitochondrial
Rabbit
Affinity chromatography
RUO
10935
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB935
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P30048
PRDX3
A synthesized peptide derived from human Peroxiredoxin 3 (aa 200-256).
20 μL