Learn More
Invitrogen™ PRDX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116556
Description
Positive Samples: U-87MG, HeLa, LO2, A-549, Mouse kidney Immunogen sequence: PAVTQHAPYF KGTAVVNGEF KDLSLDDFKG KYLVLFFYPL DFTFVCPTEI VAFSDKANEF HDVNCEVVAV SVDSHFSHLA WINTPRKNGG LGHMNIALLS DLTKQISRDY GVLLEGSGLA LRGLFIIDPN GVIKHLSVND LPVGRSVEET LRLVKAFQYV ETHGEVCPAN WTPDSPTIKP SPAASKEYFQ KVNQ.
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
Specifications
| PRDX3 | |
| Polyclonal | |
| Unconjugated | |
| PRDX3 | |
| antioxidant protein; antioxidant protein 1; anti-oxidant protein 1; Aop1; AOP-1; AW822249; D0Tohi1; Ef2l; HBC189; Mer5; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; perioredoxin-3; peroxiredoxin 3; peroxiredoxin III; Peroxiredoxin-3; Prdx3; PRO1748; Protein MER5; Protein MER5 homolog; Prx; Prx 3; PRx III; Prx3; PRX-3; Prx-III; SP22; SP-22; TDXM; thioredoxin-dependent peroxide reductase, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10935, 11757, 64371 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.01 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P20108, P30048, Q9Z0V6 | |
| PRDX3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 63-256 of human Peroxiredoxin 3 (Peroxiredoxin 3 (PRDX3)) (NP_0067841). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.