missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106901
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Peroxiredoxin 2.
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, particularly amino-terminal region containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20 - 30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
Specifications
| PRDX2 | |
| Polyclonal | |
| Unconjugated | |
| PRDX2 | |
| AL022839; Band-8; Calpromotin; epididymis secretory sperm binding protein Li 2a; HEL-S-2a; HGNC:9353; MGC4104; Natural killer cell-enhancing factor B; NkefB; NKEF-B; peroxiredoxin 2; peroxiredoxin-2; Prdx2; protector protein; PRP; Prx II-1; PRX2; PRXII; PTX1; Tdpx1; TDX1; thiol specific antioxidant protein; thiol-specific antioxidant 1; thiol-specific antioxidant protein; thioredoxin dependent peroxide reductase 1; thioredoxin peroxidase 1; thioredoxin reductase; thioredoxin-dependent peroxide reductase 1; Torin; TPx; TPX1; TPx-B; TR; Trx dependent peroxide reductase 1; TSA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21672, 29338, 7001 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32119, P35704, Q61171 | |
| PRDX2 | |
| A synthesized peptide derived from human PRDX2(Accession P32119), corresponding to amino acid residues D166-N198. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction