missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDX1 Recombinant Rabbit Monoclonal Antibody (23GB2120), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PRDX1 |
|---|---|
| Clone | 23GB2120 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088192
![]() |
Invitrogen™
MA553228 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089810
|
Invitrogen™
MA553229 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDX1 Recombinant Monoclonal antibody specifically detects PRDX1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PRDX1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P35700, Q06830 | |
| 18477, 5052 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2120 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ACR1; Alu co-repressor 1; AOEB166; B166; HBP23; heme-binding 23 kDa protein; macrophage 23 Kd stress protein; macrophage 23 kDa stress protein; macrophage 23kDa stress protein; macrophase stress protein 22kDa; macrophase stress protein 23 kd; Msp23; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NkefA; NKEF-A; OSF3; OSF-3; osteoblast specific factor 3; osteoblast-specific factor 3; PAG; Paga; PAGB; peroxiredoxin 1; peroxiredoxin-1; PRDX1; PrdxI; proliferation-associated gene A; proliferation-associated gene protein; Prx; Prx 1; PRX1; PRXI; TDPX2; TDX2; thioredoxin dependent peroxide reductase 2; thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; TPxA; Trx dependent peroxide reductase 2 | |
| A synthesized peptide derived from human PRDX1 (1-52AA). | |
| Primary | |
| -20°C | |
| PRDX1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title