Learn More
Invitrogen™ PRDX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA3750
Description
PA3-750 detects Peroxiredoxin 1 protein in human, rat and mouse samples. This antibody is specific for Prx 1 and shows no cross-reactivity with other Prx isoforms. PA3-750 has successfully been used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~26 kDa protein representing Prx 1 in human PC3 cell lysate. The PA3-750 immunizing peptide corresponds to amino acid residues 103-114 from human Prx 1.
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
Specifications
| PRDX1 | |
| Polyclonal | |
| Unconjugated | |
| PRDX1 | |
| ACR1; Alu co-repressor 1; AOEB166; B166; HBP23; heme-binding 23 kDa protein; macrophage 23 Kd stress protein; macrophage 23 kDa stress protein; macrophage 23kDa stress protein; macrophase stress protein 22kDa; macrophase stress protein 23 kd; Msp23; Natural killer cell-enhancing factor A; natural killer-enhancing factor A; NkefA; NKEF-A; OSF3; OSF-3; osteoblast specific factor 3; osteoblast-specific factor 3; PAG; Paga; PAGB; peroxiredoxin 1; peroxiredoxin-1; PRDX1; PrdxI; proliferation-associated gene A; proliferation-associated gene protein; Prx; Prx 1; PRX1; PRXI; TDPX2; TDX2; thioredoxin dependent peroxide reductase 2; thioredoxin peroxidase 2; Thioredoxin-dependent peroxide reductase 2; TPxA; Trx dependent peroxide reductase 2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P35700, Q06830, Q63716 | |
| PRDX1 | |
| Synthetic peptide corresponding to residues L(103) V S D P K R T I A Q D(114) of human Peroxiredoxin 1. | |
| RUO | |
| 117254, 18477, 5052 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.