missing translation for 'onlineSavingsMsg'
Learn More

PRDX1 Mouse anti-Human, Mouse, Rat, Clone: 2B2A2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66820-1-IG-20UL

 View more versions of this product

Product Code. 16889045

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.
Specifications

Specifications

PRDX1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P35700, Q06830, Q63716
Prdx1
PRDX1 Fusion Protein Ag8821
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG1
Western Blot
2B2A2
Unconjugated
PRDX1
MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2
Mouse
Protein G
RUO
117254, 18477, 5052
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.