missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX1 Mouse anti-Human, Mouse, Rat, Clone: 2B2A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66820-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.Specifications
| PRDX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35700, Q06830, Q63716 | |
| Prdx1 | |
| PRDX1 Fusion Protein Ag8821 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2B2A2 | |
| Unconjugated | |
| PRDX1 | |
| MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 117254, 18477, 5052 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction