missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569010
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Horse: 100%; Human: 100%; Mouse: 77%; Pig: 100%; Rabbit: 100%; Rat: 77%.
This gene encodes a member of a family of proteins that may have roles in transcription and other nuclear processes. The encoded protein contains a KRAB (Kruppel-associated box) domain -A box and a SET (Su(var)3-9, Enhancer-of-zeste, Trithorax) domain and may function as a histone methyltransferase.
Specifications
| PRDM7 | |
| Polyclonal | |
| Unconjugated | |
| PRDM7 | |
| histone-lysine N-methyltransferase PRDM9-like; PFM4; PR domain 7; PR domain containing 7; PR domain containing 9; PR domain zinc finger protein 7; PR domain-containing protein 7; PR/SET domain 7; PRDM7; Prdm9; PR-domain family protein 4; probable histone-lysine N-methyltransferase PRDM7; ZNF910 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11105 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NQW5 | |
| PRDM7 | |
| synthetic peptide directed towards the N-terminal region of human PRDM7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction