missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM4 Recombinant Human Monoclonal Antibody (RAB-C367)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551850
This item is not returnable.
View return policy
Description
This antibody recognizes PRDM4 (PR domain zinc finger protein 4). It binds to a folded domain, amino acids 390-540. PRDM4 is a transcription factor involved in cell differentiation.
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.
Specifications
| PRDM4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9UKN5 | |
| PRDM4 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Immunocytochemistry, Mass Spectrometry | |
| RAB-C367 | |
| Unconjugated | |
| PRDM4 | |
| 1700031E19Rik; 2810470D21Rik; AW552272; MGC45046; PFM1; PR domain 4; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PRDM4; PR-domain zinc-finger protein PFM1; SC-1 | |
| Human | |
| Metal-chelate chromatography | |
| RUO | |
| 11108 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction