missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM4 Antibody (RAB-C367) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
£310.00 - £657.00
Specifications
| Antigen | PRDM4 |
|---|---|
| Clone | RAB-C367 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679723
|
Novus Biologicals
NBP3-20138-0.025mg |
0.025 mg |
£310.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680905
|
Novus Biologicals
NBP3-20138-0.1mg |
0.1 mg |
£657.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM4 Monoclonal antibody specifically detects PRDM4 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| PRDM4 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC45046, PFM1PR domain-containing protein 4, PR domain containing 4, PR domain zinc finger protein 4, PR-domain zinc-finger protein PFM1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM4 protein under non-denaturing conditions. | |
| Affinity purified |
| RAB-C367 | |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| Unconjugated | |
| Human | |
| Chromatin Research | |
| PBS | |
| 11108 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title