missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM2 Monoclonal Antibody (33AT1045)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA511144
This item is not returnable.
View return policy
Description
This antibody is specific for RIZ1, it does not recognize the isoform beta (RIZ2).
PRDM2, a member of nuclear/histone methyltransferase superfamily, consists of an N-terminal PR domain, an Rb-binding motif, 8 Kruppel-like zinc finger motifs, Nuclear hormone-receptor binding motif and a C-terminal PR-domain binding motif. Ubiquitously expressed in low levels, PRDM2 exists in 2 isoforms, a 280 kDa RIZ1 containing PR-domain and a 250 kDa truncated RIZ2 that lacks PR-domain. A tumor suppressor with a prominent role in cell growth and differentiation, PRDM2 binds to Retinoblastoma protein, estrogen receptor, GATA-3 and TPA-responsive element (MTE) of Heme-oxygenase-1 gene and influences pathogenesis of Retinoblastoma, transcriptional regulation during neuronal differentiation, transactivation of Heme-oxygenase-1 gene and specific effector of estrogen action. It is inactivated or altered due to mutations and hypermethylation in many types of human cancers suggesting a potential biomarker.
Specifications
| PRDM2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q13029 | |
| PRDM2 | |
| Purified recombinant GST fusion protein encoding aa 1-347 of human RIZ1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ChIP Assay, Western Blot | |
| 33AT1045 | |
| Unconjugated | |
| PRDM2 | |
| 4833427P12Rik; E330024L24Rik; G3b; GATA-3 binding protein G3B; GATA-3-binding protein G3B; Gm1033; Gm19732; HUMHOXY1; KMT8; lysine N-methyltransferase 8; MTBzf; MTB-ZF; MTE-binding protein; PR domain 2; PR domain containing 2, with ZNF domain; PR domain zinc finger protein 2; PR domain-containing protein 2; Prdm2; retinoblastoma interacting zinc finger protein; retinoblastoma protein-binding zinc finger protein; retinoblastoma protein-interacting zinc finger protein; RIZ; RIZ1; RIZ2; RP5-1177E19.1; zinc finger protein RIZ; zinc-finger DNA-binding protein; Znfpr1c1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 110593, 7799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction