missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM16 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142015
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells.Specifications
PRDM16 | |
Polyclonal | |
Unconjugated | |
PRDM16 | |
5730557K01Rik; CMD1LL; csp1; Histone-lysine N-methyltransferase PRDM16; Kiaa1675; line 27; LVNC8; MDS1/EVI1-like gene 1; MEL1; PFM13; PR domain 16; PR domain containing 16; PR domain zinc finger protein 16; PR domain-containing protein 16; Prdm16; Transcription factor MEL1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
63976, 70673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
ChIP Assay, Flow Cytometry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
A2A935, Q9HAZ2 | |
PRDM16 | |
Peptide with sequence C-TSESGAFHPINHL. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |