missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565042
This item is not returnable.
View return policy
Description
Immunogen sequence: DKPMLKQAGSG VHAAGTPENS APVESEPSQW ACKVCSATFL ELQLLNEHLL GHLEQAKSLP PGSQSEAAAP EKEQD Highest antigen sequence identity to the following orthologs - mouse 76%, rat 75%.
The exact function of PRDM15 remains unknown.
Specifications
| PRDM15 | |
| Polyclonal | |
| Unconjugated | |
| PRDM15 | |
| C21orf83; E130018M06Rik; ORF62; PFM12; PFM15; PR domain 15; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; PR/SET domain 15; PRDM15; Zfp298; Zinc finger protein 298; ZNF298 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57071 | |
| PRDM15 | |
| Recombinant Human PRDM15. Recombinant protein control fragment (Product #RP-105809). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction