missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM15 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00063977-B01P
This item is not returnable.
View return policy
Description
PRDM15 Polyclonal antibody specifically detects PRDM15 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PRDM15 | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:2000 | |
| AAH67102.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 63977 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| C21orf83, chromosome 21 open reading frame 83, PFM15, PR domain containing 15, PR domain zinc finger protein 15, PR domain-containing protein 15, Zinc finger protein 298, ZNF298 | |
| PRDM15 (AAH67102.1, 1 a.a. - 1141 a.a.) full-length human protein. MAEDGSEEIMFIWCEDCSQYHDSECPELGPVVMVKDSFVLSRARSSLPPNLEIRRLEDGAEGVFAITQLVKRTQFGPFESRRVAKWEKESAFPLKVFQKDGHPVCFDTSNEDDCNWMMLVRPAAEAEHQNLTAYQHGSDVYFTTSRDIPPGTELRVWYAAFYAKKMDKPMLKQAGSGVHAAGTPENSAPVESEPSQWACKVCSATFLELQLLNEHLLGHLEQAKSLPPGSQSEAAAPEKEQDTPRGEPPAVPESENVATKEQKKKPRRGRKPKVSKAEQPLVIVEDKEPTEQVAEIITEVPPDEPVSATPDERIMELVLGKLATTTTDTSSVPKFTHHQNNTITLKRSLILSSRHGIRRKVIKQLGEHKRVYQCNICSKIFQNSSNLSRHVRSHGDKLFKCEECAKLFSRKESLKQHVSYKHSRNEVDGEYRYRCGTCEKTFRIESALEFHNCRTDDKTFQCEMCFRFFSTNSNLSKHKKKHGDKKFACEVCSKMFYRKDVMLDHQRRHLEGVRRVKREDLEAGGENLVRYKKEPSGCPVCGKVFSCRSNMNKHLLTHGDKKYTCEICGRKFFRVDVLRDHIHVHFKDIALMDDHQREEFIGKIGISSEENDDNSDESADSEPHKYSCKRCQLTFGRGKEYLKHIMEVHKEKGYGCSICNRRFALKATYHAHMVIHRENLPDPNVQKYIHPCEICGRIFNSIGNLERHKLIHTGVKSHACEQCGKSFARKDMLKEHMRVHDNVREYLCAECGKGMKTKHALRHHMKLHKGIKEYECKECHRRFAQKVNMLKHCKRHTGIKDFMCELCGKTFSERNTMETHKLIHTVGKQWTCSVCDKKYVTEYMLQKHVQLTHDKVEAQSCQLCGTKVSTRASMSRHMRRKHPEVLAVRIDDLDHLPETTTIDASSIGIVQPELTLEQEDLAEGKHGKAAKRSHKRKQKPEEEAGAPVPEDATFSEYSEKETEFTGSVGDETNSAVQSIQQVVVTLGDPNVTTPSSSVGLTNITVTPITTAAATQFTNLQPVAVGHLTTPERQLQLDNSILTVTFDTVSGSAMLHNRQNDVQIHPQPEASNPQSVAHFINLTTLVNSITPLGSQLSDQHPLTWRAVPQTDVLPPPQPQAPPQQAAQPQVQAEQQQQQMYSY | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction