missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PRDM15 293T Cell Transient Overexpression Lysate

Product Code. 18429238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429238 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429238 Supplier Novus Biologicals™ Supplier No. H00063977T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target PRDM15
Synonym C21orf83, chromosome 21 open reading frame 83, PFM15, PR domain containing 15, PR domain zinc finger protein 15, PR domain-containing protein 15, Zinc finger protein 298, ZNF298
Gene Symbol PRDM15
Product Title
Select an issue

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.