missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144977
This item is not returnable.
View return policy
Description
Immunogen sequence: PLEWIGLIRA ARNSQEQTLE AIADLPGGQI FYRALRDVQP GEELTVWYSN SLAQWFDIPT TATPTHDEKG EERYICWYCW Antigen sequence identity: Mouse 99%; Rat 99%.
PRDM13 may be involved in transcriptional regulation.
Specifications
| PRDM13 | |
| Polyclonal | |
| Unconjugated | |
| PRDM13 | |
| MU-MB-20.220; PFM10; PR domain 13; PR domain containing 13; PR domain zinc finger protein 13; PR domain-containing protein 13; PR/SET domain 13; PRDM13; PR-domain containing protein 13; PR-domain zinc finger protein 13 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 59336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4Q3 | |
| PRDM13 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 64-143. Recombinant protein control fragment (Product #RP-110127). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction