missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £460.00
Specifications
| Antigen | PRDM12 |
|---|---|
| Dilution | Western Blot 0.4 ul/ml, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18446360
|
Novus Biologicals
NBP1-85829-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18248377
|
Novus Biologicals
NBP1-85829 |
0.1 mL |
£460.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRDM12 Polyclonal specifically detects PRDM12 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PRDM12 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 59335 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MWEVFNEDGTVRYFIDASQEDHRSWMTYIKCARNEQEQNLEVVQIGTSIFYKAIEMIPPDQELLVWYGNSHNTF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ul/ml, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| PFM9, PR domain containing 12, PR domain zinc finger protein 12, PR domain-containing protein 12, PR-domain containing protein 12, PR-domain zinc finger protein 12 | |
| PRDM12 | |
| IgG | |
| Affinity Purified | |
| Specificity of human PRDM12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title