missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584328
This item is not returnable.
View return policy
Description
Immunogen sequence: QVDFWFCESC QEYFVDECPN HGPPVFVSDT PVPVGIPDRA ALTIPQGMEV VKDTSGESDV RCVNEVIPKG HIFGPYEGQI STQD Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 93%.
PRDM11 (PR/SET Domain 11) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and methyltransferase activity. May be involved in transcription regulation.
Specifications
| PRDM11 | |
| Polyclonal | |
| Unconjugated | |
| PRDM11 | |
| 8030443D09Rik; AA163972; Gm707; PFM8; PR domain 11; PR domain containing 11; PR domain-containing protein 11; PRDM11; PR-domain containing protein 11; RGD1305673 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56981 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQV5 | |
| PRDM11 | |
| Recombinant protein corresponding to Human PRDM11. Recombinant protein control fragment (Product #RP-101714). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction