missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRDM10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104031
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRDM10.
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.
Specifications
| PRDM10 | |
| Polyclonal | |
| Unconjugated | |
| PRDM10 | |
| Gm1112; KIAA1231; MGC131802; PFM7; PR domain 10; PR domain containing 10; PR domain zinc finger protein 10; PR domain-containing protein 10; PRDM zinc finger transcription factor; PRDM10; PR-domain family member 7; RGD1560384; TRIS; tristanin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 382066, 500964, 56980 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UTQ7, Q9NQV6 | |
| PRDM10 | |
| A synthesized peptide derived from human PRDM10(Accession Q9NQV6), corresponding to amino acid residues V981-Q1031. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction