missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDC/GREM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57598
This item is not returnable.
View return policy
Description
PRDC/GREM2 Polyclonal specifically detects PRDC/GREM2 in Human samples. It is validated for Western Blot.
Specifications
| PRDC/GREM2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CKTSF1B2FLJ21195, Cysteine knot superfamily 1, BMP antagonist 2, DAN domain family member 3, DAND3Prdc, gremlin 2, gremlin 2, cysteine knot superfamily, homolog, gremlin-2, PRDCgremlin 2, cysteine knot superfamily, homolog (Xenopus laevis), Protein related to DAN and cerberus | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H772 | |
| GREM2 | |
| Synthetic peptides corresponding to GREM2 (gremlin 2, cysteine knot superfamily, homolog (Xenopus laevis)) The peptide sequence was selected from the N terminal of GREM2)(50ug). Peptide sequence MFWKLSLSLFLVAVLVKVAEARKNRPAGAIPSPYKDGSSNNSERWQHQIK The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Stem Cell Markers | |
| 64388 | |
| Human, Mouse, Rat, Canine, Equine, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction