missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRCC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553993
This item is not returnable.
View return policy
Description
Immunogen sequence: RNRGREEINF VEIKGDDQLS GAQQWMTKSL TEEKTMKSFS KKKGEQPTGQ QRRKHQITYL IHQAKERELE LKNTWSENKL SRRQTQAKY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PRCC may regulate cell cycle progression through interaction with MAD2L2.
Specifications
| PRCC | |
| Polyclonal | |
| Unconjugated | |
| Prcc | |
| LOW QUALITY PROTEIN: proline-rich protein PRCC; papillary renal cell carcinoma (translocation-associated); papillary renal cell carcinoma translocation-associated gene product; papillary renal cell carcinoma translocation-associated gene protein; PRCC; PRCC, proline rich mitotic checkpoint control factor; proline rich mitotic checkpoint control factor; proline-rich protein PRCC; RCCP1; si:ch211-210h11.8; TPRC; Unknown (protein for MGC:140094) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5546 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92733 | |
| Prcc | |
| Recombinant protein corresponding to Human PRCC. Recombinant protein control fragment (Product #RP-92570). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction