missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15617-1-AP-150UL
This item is not returnable.
View return policy
Description
The dual bands between 66-72 kDa detected by this antibody may represent different forms of PRC1.
This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.Specifications
| PRC1 | |
| Polyclonal | |
| Unconjugated | |
| PRC1 | |
| ASE1, PRC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9055 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43663 | |
| PRC1 | |
| PRC1 Fusion Protein Ag8028 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction