missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRC1 Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 1E1D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67027-100UL
This item is not returnable.
View return policy
Description
PRC1 Monoclonal antibody specifically detects PRC1 in Human, Mouse, Rat samples. It is validated for IFSpecifications
| PRC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 100 μL | |
| 9055 | |
| Store at -20°C | |
| Liquid |
| Immunofluorescence | |
| 1E1D5 | |
| CoraLite 488 | |
| BC111712 | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction